missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethoxybenzoic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC115450250

 View more versions of this product

Catalog No. AC115450250



Description

Description

Specifications

Specifications

3, 4-Dimethoxybenzoic acid
Authentic
0.5% max. (K.F.)
179°C to 182°C
93-07-2
100.0
C9H10O4
MFCD00002500
15, 10147
Solubility in water: 0.5g/L (14°C). Other solubilities: readily soluble in alcohol,soluble in ether
COC1=C(C=C(C=C1)C(=O)O)OC
182.18
CHEBI:296881
Crystalline Powder
>100°C
Glass bottle
Beige to White or Red
25g
99.0
99% min. (HPLC)
(CH3O)2C6H3CO2H
10, I, 188
3,4-dimethoxy-benzoic acid, 3,4-dimethoxybenzoes, 3,4-dimethylprotocatechuic acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, unii-1yy04e7rr4, veratric acid, veratrumenoic acid, veratrylic acid
DAUAQNGYDSHRET-UHFFFAOYSA-N
3,4-dimethoxybenzoic acid
7121
182.18
99+%

Chemical Identifiers

93-07-2
182.18
DAUAQNGYDSHRET-UHFFFAOYSA-N
7121
3,4-dimethoxybenzoic acid
C9H10O4
MFCD00002500
3,4-dimethoxy-benzoic acid, 3,4-dimethoxybenzoes, 3,4-dimethylprotocatechuic acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, unii-1yy04e7rr4, veratric acid, veratrumenoic acid, veratrylic acid
CHEBI:296881
COC1=C(C=C(C=C1)C(=O)O)OC
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 202-215-7

SDS Documents