missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxybenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1526314

 View more versions of this product

Catalog No. AAA1526314



Specifications

Specifications

3,4-Dimethoxybenzenesulfonyl chloride
25g
C8H9ClO4S
UN3261
3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
236.666
236.68
66°C to 70°C
23095-31-0
MFCD00051769
2840170
RSJSYCZYQNJQPY-UHFFFAOYSA-N
3,4-dimethoxybenzenesulfonyl chloride
2734183
98%

Chemical Identifiers

23095-31-0
236.666
RSJSYCZYQNJQPY-UHFFFAOYSA-N
2734183
COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC
C8H9ClO4S
MFCD00051769
3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
3,4-dimethoxybenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents