Learn More
3,4-Dimethoxy-N-methylbenzylamine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439680010
Specifications
3, 4-Dimethoxy-N-methylbenzylamine | |
Authentic | |
Brown to Colorless | |
63-64-9 | |
100.0 | |
C10H15NO2 | |
XQODFBIAQVJQHF-UHFFFAOYSA-N | |
1-(3,4-dimethoxyphenyl)-N-methylmethanamine | |
592161 | |
Liquid |
110°C | |
Glass Bottle | |
1g | |
96.0 | |
96% min. (GC) | |
1-3,4-dimethoxyphenyl-n-methyl-methanamine, 1-3,4-dimethoxyphenyl-n-methylmethanamine, 3,4-dimethoxy-benzyl-methyl-amine, 3,4-dimethoxy-n-methylbenzylamine, 3,4-dimethoxybenzyl methylamine, 3,4-dimethoxybenzyl-n-methylamine, 3,4-dimethoxyphenyl methyl methyl amine, 3,4-dimethoxyphenyl methyl methylamine, benzenemethanamine, 3,4-dimethoxy-n-methyl, n-methyl-3,4-dimethoxybenzylamine | |
CNCC1=CC(=C(C=C1)OC)OC | |
181.24 | |
181.24 | |
97% |
Chemical Identifiers
63-64-9 | |
181.24 | |
1-3,4-dimethoxyphenyl-n-methyl-methanamine, 1-3,4-dimethoxyphenyl-n-methylmethanamine, 3,4-dimethoxy-benzyl-methyl-amine, 3,4-dimethoxy-n-methylbenzylamine, 3,4-dimethoxybenzyl methylamine, 3,4-dimethoxybenzyl-n-methylamine, 3,4-dimethoxyphenyl methyl methyl amine, 3,4-dimethoxyphenyl methyl methylamine, benzenemethanamine, 3,4-dimethoxy-n-methyl, n-methyl-3,4-dimethoxybenzylamine | |
1-(3,4-dimethoxyphenyl)-N-methylmethanamine |
C10H15NO2 | |
XQODFBIAQVJQHF-UHFFFAOYSA-N | |
592161 | |
CNCC1=CC(=C(C=C1)OC)OC |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger