missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxy-L-phenylalanine, 97%

Manufacturer:  Alfa Aesar™ H5336814

 View more versions of this product

Catalog No. AAH5336814



Specifications

Specifications

3,4-Dimethoxy-L-phenylalanine
C11H15NO4
3,4-dimethoxy-l-phenylalanine, s-2-amino-3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl-l-alanine, l-3,4-dimethoxyphenylalanine, l-3,4-dimethoxyphenyl alanine, tyrosine, 3-methoxy-o-methyl, 3,4-dimethoxyphenylalanine, l, unii-gc6uei64ec, s-2-amino-3-3,4-dimethoxy-phenyl-propionic acid, l-tyrosine, 3-methoxy-o-methyl
COC1=C(C=C(C=C1)CC(C(=O)O)N)OC
225.244
225.24
254°C to 257°C
−5° (c=4 in 1M Hydrochloric Acid)
32161-30-1
MFCD01321274
VWTFNYVAFGYEKI-QMMMGPOBSA-N
(2S)-2-amino-3-(3,4-dimethoxyphenyl)propanoic acid
10632946
97%
25g

Chemical Identifiers

32161-30-1
225.244
VWTFNYVAFGYEKI-QMMMGPOBSA-N
10632946
COC1=C(C=C(C=C1)CC(C(=O)O)N)OC
C11H15NO4
MFCD01321274
3,4-dimethoxy-l-phenylalanine, s-2-amino-3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl-l-alanine, l-3,4-dimethoxyphenylalanine, l-3,4-dimethoxyphenyl alanine, tyrosine, 3-methoxy-o-methyl, 3,4-dimethoxyphenylalanine, l, unii-gc6uei64ec, s-2-amino-3-3,4-dimethoxy-phenyl-propionic acid, l-tyrosine, 3-methoxy-o-methyl
(2S)-2-amino-3-(3,4-dimethoxyphenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents