Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxy-beta-nitrostyrene, 99%

$60.66 - $618.80

Chemical Identifiers

Chemical Name or Material 3,4-Dimethoxy-beta-nitrostyrene
CAS 4230-93-7
Molecular Formula C10H11NO4
Formula Weight 209.19
MDL Number MFCD00024823
Synonym 1-(3,4-Dimethoxyphenyl)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2513906

  SDS
alfa aesar™
B2513906
5g Each for $60.66

AAB2513914

  SDS
alfa aesar™
B2513914
25g Each for $198.57

AAB2513922

  SDS
alfa aesar™
B2513922
100g Each for $618.80
Description & Specifications

Specifications

Formula Weight 209.19
Chemical Name or Material 3,4-Dimethoxy-beta-nitrostyrene
Assay 99%
Molecular Formula C10H11NO4
Melting Point 139° to 141°C
MDL Number MFCD00024823
CAS 4230-93-7
Quantity 5g
Synonym 1-(3,4-Dimethoxyphenyl)-2-nitroethylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.