missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimercaptotoluene 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 421180050

 View more versions of this product

Catalog No. AC421180050


 

Specifications Description & Specifications

Specifications

3, 4-Dimercaptotoluene
496-74-2
100.0
Glass bottle
1.1800g/mL
95%
White to yellow
MFCD00004844
15, 9690
95%
94.0
Authentic
Crystalline Low Melting Solid
≥94%
C7H8S2
156.27
5g
3, 4-Toluenedithiol

Chemical Identifiers

496-74-2
100.0
C7H8S2
MFCD00004844
3, 4-Dimercaptotoluene, 95%
94.0
95%
156.27
3, 4-Toluenedithiol
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-828-3

RTECSNumber : XT2275000

TSCA : TSCA