Please login to your online account to display your discounted pricing

3,4-Dimercaptotoluene 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  421180010

 View more versions of this product

Catalog No. AC421180010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Low Melting Solid
Formula Weight 156.27
Quantity 1g
Color White to yellow
EINECS Number 207-828-3
TSCA TSCA
Density 1.1800g/mL
RTECS Number XT2275000
Merck Index 15, 9690
CAS Min % 94.0
CAS Max % 100.0
Synonym 3, 4-Toluenedithiol
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Chemical Name or Material 3, 4-Dimercaptotoluene
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C7H8S2
MDL Number MFCD00004844
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%
CAS 496-74-2

Specifications

Chemical Name or Material 3, 4-Dimercaptotoluene
Name Note 95%
CAS 496-74-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H8S2
Formula Weight 156.27
MDL Number MFCD00004844
Synonym 3, 4-Toluenedithiol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents