Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4'-Diisopropylbiphenyl, 94%

$25.62 - $94.93

Chemical Identifiers

Chemical Name or Material 3,4'-Diisopropylbiphenyl
CAS 61434-46-6
Molecular Formula C18H22
Formula Weight 238.37
MDL Number MFCD00043539
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2739306

  SDS
alfa aesar™
H2739306
5g Each for $25.62

AAH2739314

  SDS
alfa aesar™
H2739314
25g Each for $40.69

AAH2739322

  SDS
alfa aesar™
H2739322
100g Each for $94.93
Description & Specifications

Specifications

Formula Weight 238.37
Assay 94%
Chemical Name or Material 3,4'-Diisopropylbiphenyl
CAS 61434-46-6
Quantity 5g
MDL Number MFCD00043539
Molecular Formula C18H22