Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4'-Dihydroxyflavone, 97%

$88.15 - $88.15

Chemical Identifiers

Chemical Name or Material 3',4'-Dihydroxyflavone
CAS 4143-64-0
Molecular Formula C15H10O4
Formula Weight 254.24
MDL Number MFCD00017600
Beilstein 212540
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL14161MC

  SDS
alfa aesar™
L14161MC
100mg Each for $88.15
Description & Specifications

Specifications

Formula Weight 254.24
CAS 4143-64-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3',4'-Dihydroxyflavone
Molecular Formula C15H10O4
Beilstein 212540
Health Hazard 1 Warning
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100mg
Melting Point 249° to 252°C
MDL Number MFCD00017600
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.