Learn More
3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114930250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 4-Dihydroxycinnamic acid | |
99.0 | |
194.0°C to 198.0°C | |
Authentic | |
Glass bottle | |
(HO)2C6H3CH=CHCO2H | |
10,436 | |
caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid | |
QAIPRVGONGVQAS-DUXPYHPUSA-N | |
(E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid | |
689043 | |
180.16 | |
Powder |
331-39-5 | |
100.0 | |
Green-Yellow to Yellow | |
99+% | |
C9H8O4 | |
25 g | |
15,1638 | |
Solubility in water: soluble in hot water. Other solubilities: freely soluble in cold alcohol | |
C1=CC(=C(C=C1C=CC(=O)O)O)O | |
180.16 | |
CHEBI:16433 | |
99+% |
Chemical Identifiers
331-39-5 | |
180.16 | |
caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid | |
CHEBI:16433 | |
C1=CC(=C(C=C1C=CC(=O)O)O)O |
C9H8O4 | |
QAIPRVGONGVQAS-DUXPYHPUSA-N | |
689043 | |
(E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of damaging fertility or the unborn child.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 206-361-2
RUO – Research Use Only