missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydroxybenzylamine hydrobromide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 228570050

 View more versions of this product

Catalog No. AC228570050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Dihydroxybenzylamine hydrobromide
Glass bottle
>180.0°C
98%
97.5
98%
(HO)2C6H3CH2NH2·HBr
13, 796
BVFZTXFCZAXSHN-UHFFFAOYSA-N
4-(aminomethyl)benzene-1,2-diol;hydrobromide
13343562
Crystalline Powder
Authentic
Beige
5g
16290-26-9
100.0
C7H9NO2·HBr
MFCD00012859
1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide
C1=CC(=C(C=C1CN)O)O.Br
220.07
220.07
98%

Chemical Identifiers

16290-26-9
220.07
BVFZTXFCZAXSHN-UHFFFAOYSA-N
13343562
C1=CC(=C(C=C1CN)O)O.Br
C7H9NO2·HBr
MFCD00012859
1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide
4-(aminomethyl)benzene-1,2-diol;hydrobromide
Safety and Handling

Safety and Handling

EINECSNumber : 240-382-8

TSCA : TSCA

SDS Documents