missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydroxybenzylamine hydrobromide, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 228570010
Specifications
3, 4-Dihydroxybenzylamine hydrobromide | |
Glass bottle | |
>180.0°C | |
98% | |
97.5 | |
98% | |
(HO)2C6H3CH2NH2·HBr | |
13, 796 | |
BVFZTXFCZAXSHN-UHFFFAOYSA-N | |
4-(aminomethyl)benzene-1,2-diol;hydrobromide | |
13343562 | |
Crystalline Powder |
Authentic | |
Beige | |
1g | |
16290-26-9 | |
100.0 | |
C7H9NO2·HBr | |
MFCD00012859 | |
1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide | |
C1=CC(=C(C=C1CN)O)O.Br | |
220.07 | |
220.07 | |
98% |
Chemical Identifiers
16290-26-9 | |
220.07 | |
BVFZTXFCZAXSHN-UHFFFAOYSA-N | |
13343562 | |
C1=CC(=C(C=C1CN)O)O.Br |
C7H9NO2·HBr | |
MFCD00012859 | |
1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide | |
4-(aminomethyl)benzene-1,2-diol;hydrobromide |
Safety and Handling
EINECSNumber : 240-382-8
TSCA : TSCA