missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydroxybenzylamine hydrobromide, 98%, ACROS Organics™

$48.74 - $99.50

Chemical Identifiers

CAS 16290-26-9
Molecular Formula C7H9NO2·HBr
Molecular Weight (g/mol) 220.07
MDL Number MFCD00012859
InChI Key BVFZTXFCZAXSHN-UHFFFAOYSA-N
Synonym 1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide
PubChem CID 13343562
IUPAC Name 4-(aminomethyl)benzene-1,2-diol;hydrobromide
SMILES C1=CC(=C(C=C1CN)O)O.Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC228572500
SDS View Documents
Thermo Fisher Scientific
228572500
250mg Glass bottle
Each for $48.74
 
AC228570010
SDS View Documents
Fisher Scientific Acros
228570010
1g Glass bottle
Each for $99.50
 
AC228570050
SDS View Documents
Thermo Fisher Scientific
228570050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

16290-26-9
220.07
BVFZTXFCZAXSHN-UHFFFAOYSA-N
13343562
C1=CC(=C(C=C1CN)O)O.Br
C7H9NO2·HBr
MFCD00012859
1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide
4-(aminomethyl)benzene-1,2-diol;hydrobromide

Specifications

Authentic
Beige
250mg
97.5
98%
(HO)2C6H3CH2NH2·HBr
13, 796
BVFZTXFCZAXSHN-UHFFFAOYSA-N
4-(aminomethyl)benzene-1,2-diol;hydrobromide
13343562
Crystalline Powder
3, 4-Dihydroxybenzylamine hydrobromide, 98%
Glass bottle
>180.0°C
16290-26-9
100.0
C7H9NO2·HBr
MFCD00012859
1, 4-aminomethyl-, hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide, 4-aminomethyl catechol hydrobromide, acmc-1afa4, dhba hydrobromide
C1=CC(=C(C=C1CN)O)O.Br
220.07
220.07
98%
Safety and Handling

Safety and Handling

EINECSNumber : 240-382-8

TSCA : TSCA

SDS