missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydroxybenzonitrile, 97%

Manufacturer:  Alfa Aesar™ A1478303

 View more versions of this product

Catalog No. AAA1478303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3,4-Dihydroxybenzonitrile
1g
C7H5NO2
UN3439
protocatachuonitrile, 3,4-dihydroxy benzonitrile, 3,4-dihydroxybenzenecarbonitrile, protocatechuonitrile, 4-cyanocatechol, protocatechunitrile, 4-cyanopyrocatechol, pubchem3120, acmc-209e6q, 3, 4-dihydroxybenzonitrile
C1=CC(=C(C=C1C#N)O)O
135.122
135.12
153°C to 156°C (decomposition)
17345-61-8
MFCD00016436
2082204
NUWHYWYSMAPBHK-UHFFFAOYSA-N
3,4-dihydroxybenzonitrile
87065
97%

Chemical Identifiers

17345-61-8
135.122
NUWHYWYSMAPBHK-UHFFFAOYSA-N
87065
C1=CC(=C(C=C1C#N)O)O
C7H5NO2
MFCD00016436
protocatachuonitrile, 3,4-dihydroxy benzonitrile, 3,4-dihydroxybenzenecarbonitrile, protocatechuonitrile, 4-cyanocatechol, protocatechunitrile, 4-cyanopyrocatechol, pubchem3120, acmc-209e6q, 3, 4-dihydroxybenzonitrile
3,4-dihydroxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents