Learn More
3,4-Dihydroxybenzoic acid, 97%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 114891000
Specifications
3, 4-Dihydroxybenzoic acid | |
Glass bottle | |
198.0°C to 202.0°C | |
97% | |
96.0 | |
97% | |
(HO)2C6H3CO2H | |
10,389 | |
3,4-dihydroxybenzoicacid, 4,5-dihydroxybenzoic acid, 4-carboxy-1,2-dihydroxybenzene, benzoic acid, 3,4-dihydroxy, ccris 6291, chembl37537, protocatechuate, protocatechuic acid, protocatehuic acid, unii-36r5qj8l4b | |
YQUVCSBJEUQKSH-UHFFFAOYSA-N | |
3,4-dihydroxybenzoic acid | |
72 | |
154.12 | |
97% |
Authentic | |
Beige to Brown | |
100g | |
99-50-3 | |
100.0 | |
C7H6O4 | |
MFCD00002509 | |
15,8003 | |
Solubility in water: .. Other solubilities: soluble in alcohol and ether | |
C1=CC(=C(C=C1C(=O)O)O)O | |
154.12 | |
CHEBI:36062 | |
Fine Crystalline Powder |
Chemical Identifiers
99-50-3 | |
154.12 | |
YQUVCSBJEUQKSH-UHFFFAOYSA-N | |
72 | |
3,4-dihydroxybenzoic acid |
C7H6O4 | |
MFCD00002509 | |
3,4-dihydroxybenzoicacid, 4,5-dihydroxybenzoic acid, 4-carboxy-1,2-dihydroxybenzene, benzoic acid, 3,4-dihydroxy, ccris 6291, chembl37537, protocatechuate, protocatechuic acid, protocatehuic acid, unii-36r5qj8l4b | |
CHEBI:36062 | |
C1=CC(=C(C=C1C(=O)O)O)O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-760-0
RTECSNumber : UL0560000
TSCA : TSCA