Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydroxybenzaldoxime, 98%

$84.82 - $281.77

Chemical Identifiers

Chemical Name or Material 3,4-Dihydroxybenzaldoxime
CAS 3343-59-7
Molecular Formula C7H7NO3
Formula Weight 153.14
MDL Number MFCD01070622
Beilstein 3126402
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1776606

  SDS
alfa aesar™
L1776606
5g Each for $84.82

AAL1776614

  SDS
alfa aesar™
L1776614
25g Each for $281.77
Description & Specifications

Specifications

Formula Weight 153.14
MDL Number MFCD01070622
Chemical Name or Material 3,4-Dihydroxybenzaldoxime
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 3343-59-7
Molecular Formula C7H7NO3
Assay 98%
Melting Point 155° to 157°C
Beilstein 3126402
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.