missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydroxybenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1155806

 View more versions of this product

Catalog No. AAA1155806


 

Description

Description

Applications
3,4-Dihydroxybenzaldehyde is used in as an apoptosis inducer of human leukemia cells.

Solubility
Soluble in water(6.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

3,4-Dihydroxybenzaldehyde
5g
98%
MFCD00003370
14,7893
IBGBGRVKPALMCQ-UHFFFAOYSA-N
3,4-dihydroxybenzaldehyde
8768
138.12
152°C to 157°C
139-85-5
C7H6O3
774381
Protocatechualdehyde
C1=CC(=C(C=C1C=O)O)O
138.122
CHEBI:50205

Chemical Identifiers

139-85-5
138.122
IBGBGRVKPALMCQ-UHFFFAOYSA-N
8768
C7H6O3
MFCD00003370
Protocatechualdehyde
CHEBI:50205
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UL0380000

SDS Documents