missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydroxybenzaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 114851000

 View more versions of this product

Catalog No. AC114851000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Dihydroxybenzaldehyde
Glass bottle
152.0°C to 157.0°C
97%
96.0
96% min. (HPLC)
(HO)2C6H3CHO
08,246
1,2-dihydroxy-4-formylbenzene, 3,4-dihydroxy-benzaldehyde, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-benzenediol, 4-formyl-1,2-dihydroxybenzene, benzaldehyde, 3,4-dihydroxy, protocatechualdehyde, protocatechuic aldehyde, rancinamycin iv, unii-4pvp2hch4t
IBGBGRVKPALMCQ-UHFFFAOYSA-N
3,4-dihydroxybenzaldehyde
8768
138.12
97%
Authentic
Beige to Brown-Gray
100g
139-85-5
100.0
C7H6O3
MFCD00003370
15,8002
Solubility in water: 50g/L (20°C). Other solubilities: 790g/L ethanol (78°C),freely soluble in ether
C1=CC(=C(C=C1C=O)O)O
138.12
CHEBI:50205
Crystalline Powder

Chemical Identifiers

139-85-5
138.12
IBGBGRVKPALMCQ-UHFFFAOYSA-N
8768
3,4-dihydroxybenzaldehyde
C7H6O3
MFCD00003370
1,2-dihydroxy-4-formylbenzene, 3,4-dihydroxy-benzaldehyde, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-benzenediol, 4-formyl-1,2-dihydroxybenzene, benzaldehyde, 3,4-dihydroxy, protocatechualdehyde, protocatechuic aldehyde, rancinamycin iv, unii-4pvp2hch4t
CHEBI:50205
C1=CC(=C(C=C1C=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 205-377-7

RTECSNumber : UL0380000

TSCA : TSCA

SDS Documents
Product Certifications