missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydroxybenzaldehyde, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114850250
Specifications
3, 4-Dihydroxybenzaldehyde | |
Glass bottle | |
152.0°C to 157.0°C | |
97% | |
96.0 | |
96% min. (HPLC) | |
(HO)2C6H3CHO | |
08,246 | |
1,2-dihydroxy-4-formylbenzene, 3,4-dihydroxy-benzaldehyde, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-benzenediol, 4-formyl-1,2-dihydroxybenzene, benzaldehyde, 3,4-dihydroxy, protocatechualdehyde, protocatechuic aldehyde, rancinamycin iv, unii-4pvp2hch4t | |
IBGBGRVKPALMCQ-UHFFFAOYSA-N | |
3,4-dihydroxybenzaldehyde | |
8768 | |
138.12 | |
97% |
Authentic | |
Beige to Brown-Gray | |
25g | |
139-85-5 | |
100.0 | |
C7H6O3 | |
MFCD00003370 | |
15,8002 | |
Solubility in water: 50g/L (20°C). Other solubilities: 790g/L ethanol (78°C),freely soluble in ether | |
C1=CC(=C(C=C1C=O)O)O | |
138.12 | |
CHEBI:50205 | |
Crystalline Powder |
Chemical Identifiers
139-85-5 | |
138.12 | |
IBGBGRVKPALMCQ-UHFFFAOYSA-N | |
8768 | |
3,4-dihydroxybenzaldehyde |
C7H6O3 | |
MFCD00003370 | |
1,2-dihydroxy-4-formylbenzene, 3,4-dihydroxy-benzaldehyde, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-benzenediol, 4-formyl-1,2-dihydroxybenzene, benzaldehyde, 3,4-dihydroxy, protocatechualdehyde, protocatechuic aldehyde, rancinamycin iv, unii-4pvp2hch4t | |
CHEBI:50205 | |
C1=CC(=C(C=C1C=O)O)O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 205-377-7
RTECSNumber : UL0380000
TSCA : TSCA