missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydro-4-oxo-1,2,3-benzotriazine 98.0+%, TCI America™

$46.08 - $130.03

Chemical Identifiers

CAS 90-16-4
Molecular Formula C7H5N3O
Molecular Weight (g/mol) 147.137
MDL Number MFCD00052387
InChI Key DMSSTTLDFWKBSX-UHFFFAOYSA-N
Synonym 1,2,3-benzotriazin-4 1h-one, 1,2,3-benzotriazin-4 3h-one, 1,2,3-benzotriazin-4-ol, 3h-1,2,3-benzotriazin-4-one, 4-ketobenz-1,2,3-triazine, 4-ketobenzotriazine, benzazimide, benzazimidone, benzoketotriazine, usaf ma-2
PubChem CID 7006
IUPAC Name 1H-1,2,3-benzotriazin-4-one
SMILES C1=CC=C2C(=C1)C(=O)N=NN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
K00035G View Documents Tci America
K00035G
5g Each for $46.08
 
K000325G View Documents Tci America
K000325G
25g Each for $130.03
 
Specifications

Chemical Identifiers

90-16-4
147.137
DMSSTTLDFWKBSX-UHFFFAOYSA-N
7006
C1=CC=C2C(=C1)C(=O)N=NN2
C7H5N3O
MFCD00052387
1,2,3-benzotriazin-4 1h-one, 1,2,3-benzotriazin-4 3h-one, 1,2,3-benzotriazin-4-ol, 3h-1,2,3-benzotriazin-4-one, 4-ketobenz-1,2,3-triazine, 4-ketobenzotriazine, benzazimide, benzazimidone, benzoketotriazine, usaf ma-2
1H-1,2,3-benzotriazin-4-one

Specifications

Undesignated
5g
C7H5N3O
1,2,3-benzotriazin-4 1h-one, 1,2,3-benzotriazin-4 3h-one, 1,2,3-benzotriazin-4-ol, 3h-1,2,3-benzotriazin-4-one, 4-ketobenz-1,2,3-triazine, 4-ketobenzotriazine, benzazimide, benzazimidone, benzoketotriazine, usaf ma-2
C1=CC=C2C(=C1)C(=O)N=NN2
147.137
147.14
≥98.0% (HPLC,T)
218°C
90-16-4
MFCD00052387
DMSSTTLDFWKBSX-UHFFFAOYSA-N
1H-1,2,3-benzotriazin-4-one
7006
Crystal-Powder at 20°C
3,4-Dihydro-4-oxo-1,2,3-benzotriazine
Safety and Handling

Safety and Handling

RTECSNumber : DM0800000

TSCA : Yes