missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-pyran, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114785000
Specifications
3, 4-Dihydro-2H-pyran | |
−9°C | |
Glass bottle | |
0.92 | |
0.3% max. | |
Colorless to Yellow | |
500g | |
98.5 | |
98.5% min. (GC) | |
MFCD00006558 | |
01,256; 03,99; 05,220; 07,109; 10,147; 11,26 | |
2,3-dihydro-4h-pyran, 2,3-dihydropyran, 2h-3,4-dihydropyran, 2h-pyran, 3,4-dihydro, 2h-pyran, dihydro, 3,4-dihydropyran, 5,6-dihydro-4h-pyran, dihydro-2h-pyran, dihydropyran, dihydropyrane | |
BUDQDWGNQVEFAC-UHFFFAOYSA-N | |
3,4-dihydro-2H-pyran | |
8080 | |
Liquid |
0.9200g/mL | |
Authentic | |
1.4405 to 1.4425 | |
0.76 mPa.s (20°C) | |
87.0°C | |
-70.0°C | |
110-87-2 | |
100.0 | |
C5H8O | |
2376 | |
15, 3195 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol,slightly soluble in chloroform,soluble in most other organic solvents | |
C1CC=COC1 | |
84.12 | |
84.12 | |
99% |
Chemical Identifiers
110-87-2 | |
84.12 | |
BUDQDWGNQVEFAC-UHFFFAOYSA-N | |
8080 | |
C1CC=COC1 |
C5H8O | |
MFCD00006558 | |
2,3-dihydro-4h-pyran, 2,3-dihydropyran, 2h-3,4-dihydropyran, 2h-pyran, 3,4-dihydro, 2h-pyran, dihydro, 3,4-dihydropyran, 5,6-dihydro-4h-pyran, dihydro-2h-pyran, dihydropyran, dihydropyrane | |
3,4-dihydro-2H-pyran |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Danger
EINECSNumber : 203-810-4
TSCA : TSCA