Please login to your online account to display your discounted pricing

Alfa Aesar™ (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol, 97%

$53.11 - $444.85

Chemical Identifiers

Chemical Name or Material (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol
CAS 304669-35-0
Molecular Formula C7H14O2Si
Formula Weight 158.27
MDL Number MFCD09265116
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2850603

  SDS
alfa aesar™
H2850603
1g Each for $147.46

AAH2850606

  SDS
alfa aesar™
H2850606
5g Each for $444.85

AAH28506MD

  SDS
alfa aesar™
H28506MD
250mg Each for $53.11
Description & Specifications

Specifications

Formula Weight 158.27
Chemical Name or Material (3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol
Assay 97%
Molecular Formula C7H14O2Si
MDL Number MFCD09265116
CAS 304669-35-0
Boiling Point 54°C (0.8mmHg)
Quantity 1g
Flash Point 91°C (196°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

2° to 8°C