3,4-Dihydro-2H-pyran-5-carboxaldehyde, 97%, Acros Organics™
Manufacturer:  Acros Organics AC450380050
 View more versions of this product
Catalog No. AC450380050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
3,4-Dihydro-2H-pyran-5-carboxaldehyde
90.0°C (15.0 mmHg)
25090-33-9
5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde
C1CC(=COC1)C=O
112.128
112.13
Authentic
5g
C6H8O2
WWZVSCJTMPYTLY-UHFFFAOYSA-N
3,4-dihydro-2H-pyran-5-carbaldehyde
2736151
97%
Chemical Identifiers
25090-33-9
112.128
5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde
3,4-dihydro-2H-pyran-5-carbaldehyde
C6H8O2
WWZVSCJTMPYTLY-UHFFFAOYSA-N
2736151
C1CC(=COC1)C=O
Safety and Handling
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
SDS Documents