missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydro-2H-pyran-2-methanol, 96%

Manufacturer:  Alfa Aesar™ H5612406

 View more versions of this product

Catalog No. AAH5612406



Description

Description

Applications
It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

Notes
Incompatible with oxidizing agents, acids and heats. Store at room temperature.
Specifications

Specifications

3,4-Dihydro-2H-pyran-2-methanol
89°C (192°F)
92°C to 93°C (25mmHg)
3749-36-8
MFCD00051507
XMICBFRKICBBKD-UHFFFAOYSA-N
3,4-dihydro-2H-pyran-2-ylmethanol
95559
96%
1.101
1.478
5g
C6H10O2
2-hydroxymethyl-3,4-dihydro-2h-pyran
C1CC(OC=C1)CO
114.144
114.14

Chemical Identifiers

3749-36-8
114.144
XMICBFRKICBBKD-UHFFFAOYSA-N
95559
C1CC(OC=C1)CO
C6H10O2
MFCD00051507
2-hydroxymethyl-3,4-dihydro-2h-pyran
3,4-dihydro-2H-pyran-2-ylmethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 223-150-0

TSCA : No

Recommended Storage : Keep cold

Documents