missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-chromen-3-ylmethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC25809DA
Specifications
3, 4-Dihydro-2H-chromen-3-ylmethanol | |
96.5 | |
97% | |
C10H12O2 | |
1g | |
UPDSWAQIQZFOLF-UHFFFAOYSA-N | |
3,4-dihydro-2H-chromen-3-ylmethanol | |
2795460 | |
97% |
76727-28-1 | |
100.0 | |
Amber Glass Bottle | |
MFCD04972641 | |
chroman-3-ylmethanol, 3,4-dihydro-2h-1-benzopyran-3-ylmethanol, 3-hydroxymethylchroman, chroman-3-yl-methanol, chroman-3-ylmethan-1-ol, 3,4-dihydro-2h-1-benzopyran 3-methanol, 2h-1-benzopyran-3-methanol,3,4-dihydro, 2h-1-benzopyran-3-methanol, 3,4-dihydro | |
C1C(COC2=CC=CC=C21)CO | |
164.204 | |
164.2 |
Chemical Identifiers
76727-28-1 | |
164.204 | |
UPDSWAQIQZFOLF-UHFFFAOYSA-N | |
2795460 | |
C1C(COC2=CC=CC=C21)CO |
C10H12O2 | |
MFCD04972641 | |
chroman-3-ylmethanol, 3,4-dihydro-2h-1-benzopyran-3-ylmethanol, 3-hydroxymethylchroman, chroman-3-yl-methanol, chroman-3-ylmethan-1-ol, 3,4-dihydro-2h-1-benzopyran 3-methanol, 2h-1-benzopyran-3-methanol,3,4-dihydro, 2h-1-benzopyran-3-methanol, 3,4-dihydro | |
3,4-dihydro-2H-chromen-3-ylmethanol |