missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-1,5-benzodioxepine-7-carbaldehyde, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13204DE
Specifications
3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carbaldehyde | |
95% | |
C10H10O3 | |
2h-1,5-benzodioxepin-7-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxaldehyde, 2h-1,5-benzodioxepin-7-carboxaldehyde,3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepane-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepine-7-carboxaldehyde, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyd | |
C1COC2=C(C=C(C=C2)C=O)OC1 | |
178.187 | |
178.19 |
67869-90-3 | |
Amber Glass Bottle | |
5g | |
LCSVYSVGXQQHSI-UHFFFAOYSA-N | |
3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde | |
2776388 | |
95% |
Chemical Identifiers
67869-90-3 | |
178.187 | |
2h-1,5-benzodioxepin-7-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxaldehyde, 2h-1,5-benzodioxepin-7-carboxaldehyde,3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepane-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepine-7-carboxaldehyde, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyd | |
3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde |
C10H10O3 | |
LCSVYSVGXQQHSI-UHFFFAOYSA-N | |
2776388 | |
C1COC2=C(C=C(C=C2)C=O)OC1 |