Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydro-2H-1,5-benzodioxepin, 98%

$49.11 - $49.11

Chemical Identifiers

Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin
CAS 7216-18-4
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD01333457
Synonym 1,2-Trimethylenedioxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2737403

  SDS
alfa aesar™
H2737403
1g Each for $49.11
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin
Synonym 1,2-Trimethylenedioxybenzene
Assay 98%
Molecular Formula C9H10O2
MDL Number MFCD01333457
CAS 7216-18-4
RTECS Number DF4725200
Quantity 1g

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.