Please login to your online account to display your discounted pricing

3,4-Dihydro-2H-1,5-benzodioxepin-7-ylboronic acid, 90+%

Chemical Identifiers

Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylboronic acid
Assay Percent Range 90%
Molecular Formula C9H11BO4
Formula Weight 193.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC13212DA

 
fisher scientific company
CC13212DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC13212DE

 
fisher scientific company
CC13212DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC13212CB

 
fisher scientific company
CC13212CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.99
Quantity 1g
Packaging Amber Glass Bottle
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylboronic acid
Assay Percent Range 90%
Molecular Formula C9H11BO4
CAS 279261-89-1