missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-1,5-benzodioxepin-7-ylboronic acid, 90+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13212DE
Specifications
3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylboronic acid | |
90% | |
C9H11BO4 | |
3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl boronic acid, 3,4-dihydro-2h-1,5-benzodioxepine-7-boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl boronic acid, boronic acid,b-3,4-dihydro-2h-1,5-benzodioxepin-7-yl, 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-ylboronic acid, boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl, pubchem7845, acmc-1crzt, 3,4-dihydro-2h-1,5-benzodioxepin-7-boronic acid | |
B(C1=CC2=C(C=C1)OCCCO2)(O)O | |
193.993 | |
193.99 |
279261-89-1 | |
Amber Glass Bottle | |
5g | |
CSNCBPVLUIFJOS-UHFFFAOYSA-N | |
3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid | |
2776398 |
Chemical Identifiers
279261-89-1 | |
193.993 | |
3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl boronic acid, 3,4-dihydro-2h-1,5-benzodioxepine-7-boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl boronic acid, boronic acid,b-3,4-dihydro-2h-1,5-benzodioxepin-7-yl, 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-ylboronic acid, boronic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl, pubchem7845, acmc-1crzt, 3,4-dihydro-2h-1,5-benzodioxepin-7-boronic acid | |
3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid |
C9H11BO4 | |
CSNCBPVLUIFJOS-UHFFFAOYSA-N | |
2776398 | |
B(C1=CC2=C(C=C1)OCCCO2)(O)O |