Learn More
3,4-Dihydro-2H-1,5-benzodioxepin-7-amine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2786203
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water. High soluble in ethanol, acetone.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
3,4-Dihydro-2H-1,5-benzodioxepin-7-amine | |
79°C to 81°C | |
>110°C (230°F) | |
MFCD00218806 | |
3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine | |
FVLCICVRAPEYNX-UHFFFAOYSA-N | |
3,4-dihydro-2H-1,5-benzodioxepin-7-amine | |
2775654 | |
98% |
175136-34-2 | |
123°C to 128°C (0.2 mmHg) | |
C9H11NO2 | |
1 g | |
Soluble in water. High soluble in ethanol,acetone. | |
C1COC2=C(C=C(C=C2)N)OC1 | |
165.192 | |
165.19 |
Chemical Identifiers
175136-34-2 | |
165.192 | |
FVLCICVRAPEYNX-UHFFFAOYSA-N | |
3,4-dihydro-2H-1,5-benzodioxepin-7-amine |
C9H11NO2 | |
MFCD00218806 | |
2775654 | |
C1COC2=C(C=C(C=C2)N)OC1 |
Safety and Handling
GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only