missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydro-2H-1,5-benzodioxepin-6-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00413DA

 View more versions of this product

Catalog No. CC00413DA



Description

Description

Specifications

Specifications

3, 4-Dihydro-2H-1, 5-benzodioxepin-6-ylmethylamine hydrochloride
96.5
97%
C10H13NO2
1 g
ZXJOKQNPRXXPJT-UHFFFAOYSA-N
3,4-dihydro-2H-1,5-benzodioxepin-6-ylmethanamine
2794995
97%
499770-91-1
100.0
Amber Glass Bottle
MFCD04972615
3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
NCC1=C2OCCCOC2=CC=C1
179.22
215.68

Chemical Identifiers

499770-91-1
179.22
ZXJOKQNPRXXPJT-UHFFFAOYSA-N
2794995
C10H13NO2
MFCD04972615
3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
NCC1=C2OCCCOC2=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions