missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6229003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester | |
C14H20BNO3 | |
1 g | |
HFUHUNYUUCDCAU-UHFFFAOYSA-N | |
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine | |
54759084 | |
Solid |
1155264-46-2 | |
MFCD18073255 | |
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, acmc-2099qf, 3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine | |
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1 | |
261.13 | |
261.12 |
Chemical Identifiers
1155264-46-2 | |
261.13 | |
HFUHUNYUUCDCAU-UHFFFAOYSA-N | |
54759084 | |
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1 |
C14H20BNO3 | |
MFCD18073255 | |
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, acmc-2099qf, 3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine | |
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only