missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid, ≥97%, Maybridge

Chemical Identifiers

CAS 90563-93-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H9NO3
Formula Weight 179.17
MDL Number MFCD05865108
Synonym 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid (IUPAC)
Chemical Name or Material 3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC29701CB SDS Fisher Scientific Company
CC29701CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC29701DA SDS Fisher Scientific Company
CC29701DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

90563-93-2
100.0
C9H9NO3
MFCD05865108
3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
96.5
97%
179.17
4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid (IUPAC)
97%

Specifications

250mg
96.5
Amber Glass Bottle
C9H9NO3
MFCD05865108
3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
90563-93-2
100.0
97%
179.17
4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid (IUPAC)
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.