missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid, ≥97%, Maybridge

Manufacturer:  Fisher Scientific Company CC29701DA

 View more versions of this product

Catalog No. CC29701DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

179.17
97%
C9H9NO3
97%
Amber Glass Bottle
3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
1g
100.0
90563-93-2
96.5
4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid (IUPAC)
MFCD05865108

Chemical Identifiers

90563-93-2
100.0
C9H9NO3
MFCD05865108
3, 4-Dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
96.5
97%
179.17
4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid (IUPAC)
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.