Please login to your online account to display your discounted pricing

3,4-Dihydro-2-methoxy-2H-pyran, +99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  177625000

 View more versions of this product

Catalog No. AC177625000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 114.14
Quantity 500g
Color Clear colorless to yellow
EINECS Number 224-698-3
TSCA TSCA
Density 1.0000g/mL
RTECS Number UP7900000
CAS Min % 99.0
CAS Max % 100.0
Water 0.2% max.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Toxic if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Chemical Name or Material 3, 4-Dihydro-2-methoxy-2H-pyran
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C6H10O2
MDL Number MFCD00006560
CAS 4454-05-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4-Dihydro-2-methoxy-2H-pyran
Name Note 99+%
CAS 4454-05-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H10O2
Formula Weight 114.14
MDL Number MFCD00006560
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Toxic if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

SDS & Safety Documents