missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydro-2-methoxy-2H-pyran, +99%, ACROS Organics™

Chemical Identifiers

CAS 4454-05-1
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C6H10O2
Formula Weight 114.14
MDL Number MFCD00006560
Chemical Name or Material 3, 4-Dihydro-2-methoxy-2H-pyran, 99+%
Assay ≥99.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC177620050 SDS Thermo Fisher Scientific
177620050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC177625000 SDS Thermo Fisher Scientific
177625000
Glass bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

4454-05-1
100.0
C6H10O2
MFCD00006560
≥99.0%
99.0
99+%
114.14
3, 4-Dihydro-2-methoxy-2H-pyran, 99+%

Specifications

Liquid
5g
1.0000g/mL
100.0
1.4410 to 1.4430 (20°C, 589nm)
Authentic
4454-05-1
MFCD00006560
≥99.0%
114.14
Clear colorless to yellow
99.0
0.2% max.
Glass bottle
99+%
C6H10O2
3, 4-Dihydro-2-methoxy-2H-pyran, 99+%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Toxic if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-698-3

RTECSNumber : UP7900000

TSCA : TSCA