Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydro-2-methoxy-2H-pyran, 99%

Chemical Identifiers

Chemical Name or Material 3,4-Dihydro-2-methoxy-2H-pyran
CAS 4454-05-1
Molecular Formula C6H10O2
Formula Weight 114.14
MDL Number MFCD00006560
UN Number UN3271
Beilstein 107274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2460922

  SDS
alfa aesar™
B2460922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2460936

  SDS
alfa aesar™
B2460936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.14
Chemical Name or Material 3,4-Dihydro-2-methoxy-2H-pyran
TSCA TSCA
Assay 99%
Molecular Formula C6H10O2
MDL Number MFCD00006560
CAS 4454-05-1
UN Number UN3271
Beilstein 107274
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H301-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
RTECS Number UP7900000
Flash Point 16°C (61°F)
Boiling Point 126°C
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.