Please login to your online account to display your discounted pricing

3,4-Dihydro-2(1H)-quinolinone, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398010100

 View more versions of this product

Catalog No. AC398010100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 147.18
Quantity 10g
Color Off-white
Melting Point 164° to 168°C
Merck Index 15, 4817
CAS Max % 100.0
Chemical Name or Material 3, 4-Dihydro-2(1H)-quinolinone
Packaging Glass bottle
CAS 553-03-7
Assay Percent Range 98%
Molecular Formula C9H9NO
MDL Number MFCD00016722
CAS Min % 97.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4-Dihydro-2(1H)-quinolinone
CAS 553-03-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00016722
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

SDS & Safety Documents