Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dihydro-1H-isoquinoline-2-carboxamidine hydriodide, 98%

$47.26 - $47.26

Chemical Identifiers

Chemical Name or Material 3,4-Dihydro-1H-isoquinoline-2-carboxamidine hydriodide
Molecular Formula C10H13N3·HI
Formula Weight 303.14
MDL Number MFCD07681953
Synonym Debrisoquin hydriodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2773103

 
alfa aesar™
H2773103
1g Each for $47.26

AAH2773109

 
alfa aesar™
H2773109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 303.14
Chemical Name or Material 3,4-Dihydro-1H-isoquinoline-2-carboxamidine hydriodide
Assay 98%
Molecular Formula C10H13N3·HI
MDL Number MFCD07681953
Melting Point 152° to 154°C
Synonym Debrisoquin hydriodide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.