missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H54564MD

 View more versions of this product

Catalog No. AAH54564MD



Specifications

Specifications

3,4-Difluorothiobenzamide
317319-14-5
MFCD04036083
BVZJSQFOGOEVMF-UHFFFAOYSA-N
3,4-difluorobenzenecarbothioamide
16771464
97%
250mg
C7H5F2NS
3,4-difluoro-thiobenzamide
C1=CC(=C(C=C1C(=S)N)F)F
173.181
173.18

Chemical Identifiers

317319-14-5
173.181
BVZJSQFOGOEVMF-UHFFFAOYSA-N
16771464
C1=CC(=C(C=C1C(=S)N)F)F
C7H5F2NS
MFCD04036083
3,4-difluoro-thiobenzamide
3,4-difluorobenzenecarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents