Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4'-Difluoropropiophenone, 97%

$131.61 - $131.61

Chemical Identifiers

Chemical Name or Material 3',4'-Difluoropropiophenone
CAS 23384-72-7
Molecular Formula C9H8F2O
Formula Weight 170.16
MDL Number MFCD00015505
Beilstein 2088650
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1725706

 
alfa aesar™
A1725706
5g Each for $131.61
Description & Specifications

Specifications

Formula Weight 170.16
Chemical Name or Material 3',4'-Difluoropropiophenone
Assay 97%
Molecular Formula C9H8F2O
MDL Number MFCD00015505
CAS 23384-72-7
Beilstein 2088650
Boiling Point 64° to 65°C (1.8mmHg)
Quantity 5g