Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorophenylhydrazine hydrochloride, 98%

$67.58 - $190.86

Chemical Identifiers

Chemical Name or Material 3,4-Difluorophenylhydrazine hydrochloride
CAS 875664-54-3
Molecular Formula C6H6F2N2·HCl
Formula Weight 180.59
MDL Number MFCD03094170
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2618603

  SDS
alfa aesar™
H2618603
1g Each for $67.58

AAH2618606

  SDS
alfa aesar™
H2618606
5g Each for $190.86
Description & Specifications

Specifications

Formula Weight 180.59
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 3,4-Difluorophenylhydrazine hydrochloride
Assay 98%
Molecular Formula C6H6F2N2·HCl
MDL Number MFCD03094170
CAS 875664-54-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈230°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.