Please login to your online account to display your discounted pricing

3,4-Difluorophenyl isocyanate, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Difluorophenyl isocyanate
Assay ≥96.0%
Molecular Formula C7H3F2NO
Linear Formula F2C6H3NCO
Formula Weight 155.1
MDL Number MFCD00065711
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369525000

  SDS
thermo fisher scientific
AC369525000
Glass Bottle in Plastic Container 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC369520020

  SDS
thermo fisher scientific
AC369520020
Glass bottle 2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 155.1
Physical Form Liquid
Quantity 500mg
Color Yellow
Packaging Glass Bottle in Plastic Container
Density 1.3260g/mL
Chemical Name or Material 3, 4-Difluorophenyl isocyanate
Linear Formula F2C6H3NCO
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.
MDL Number MFCD00065711
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C7H3F2NO
CAS 42601-04-7
Recommended Storage Refrigerator +4°C
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.