Please login to your online account to display your discounted pricing

3,4-Difluorophenyl isocyanate, ACROS Organics™


Quantity 2g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369520020

 View more versions of this product

Catalog No. AC369520020

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 155.1
Quantity 2g
Color Yellow
Density 1.3260g/mL
Infrared Spectrum Authentic
Linear Formula F2C6H3NCO
MDL Number MFCD00065711
CAS 42601-04-7
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Recommended Storage Refrigerator +4°C
Chemical Name or Material 3, 4-Difluorophenyl isocyanate
Molecular Formula C7H3F2NO
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4-Difluorophenyl isocyanate
Assay ≥96.0%
Molecular Formula C7H3F2NO
Linear Formula F2C6H3NCO
Formula Weight 155.1
MDL Number MFCD00065711
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.

Refrigerator +4°C

SDS & Safety Documents