Please login to your online account to display your discounted pricing

3,4-Difluorophenol, 99%, ACROS Organics™

$59.42 - $59.42

Chemical Identifiers

Chemical Name or Material 3, 4-Difluorophenol
CAS 2713-33-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4F2O
Linear Formula F2(C6H3)OH
Formula Weight 130.09
MDL Number MFCD00010315
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC241960050

  SDS
thermo fisher scientific
241960050
Glass bottle 5g Each for $59.42
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 130.09
Quantity 5g
Color White to yellow
Beilstein 06, IV, 780
Melting Point 32° to 36°C
CAS Max % 100.0
CAS 2713-33-9
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 99%
Chemical Name or Material 3, 4-Difluorophenol
Molecular Formula C6H4F2O
MDL Number MFCD00010315
CAS Min % 98.5
Packaging Glass bottle
Linear Formula F2(C6H3)OH
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Flammable solid.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.