Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoromandelic acid, 97%

$38.18 - $103.76

Chemical Identifiers

Chemical Name or Material 3,4-Difluoromandelic acid
CAS 132741-29-8
Molecular Formula C8H6F2O3
Formula Weight 188.13
MDL Number MFCD00061299
Beilstein 9123299
Synonym alpha-Hydroxy-3,4-difluorophenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1098203

 
alfa aesar™
L1098203
1g Each for $103.76

AAL10982MD

 
alfa aesar™
L10982MD
250mg Each for $38.18
Description & Specifications

Specifications

Formula Weight 188.13
Assay 97%
Molecular Formula C8H6F2O3
MDL Number MFCD00061299
Beilstein 9123299
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3,4-Difluoromandelic acid
CAS 132741-29-8
Synonym alpha-Hydroxy-3,4-difluorophenylacetic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 92° to 96°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.