missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoromandelic acid, 97%

Manufacturer:  Alfa Aesar™ L1098203

 View more versions of this product

Catalog No. AAL1098203



Specifications

Specifications

3,4-Difluoromandelic acid
1g
C8H6F2O3
9123299
BKHXODARAOCNDJ-UHFFFAOYSA-N
2-(3,4-difluorophenyl)-2-hydroxyacetic acid
588647
97%
92°C to 96°C
132741-29-8
MFCD00061299
3,4-difluoromandelic acid, 2-3,4-difluorophenyl-2-hydroxyacetic acid, 3,4-difluorophenyl hydroxy acetic acid, difluoromandelicacid, 3,4-difluoro mandelic acid, 3,4-difluorophenyl hydroxy acetic acid #, alpha-hydroxy-3,4-difluorobenzeneacetic acid, 2-3,4-difluorophenyl-2-hydroxy-acetic acid, 2-3,4-bis fluoranyl phenyl-2-oxidanyl-ethanoic acid
C1=CC(=C(C=C1C(C(=O)O)O)F)F
188.13
188.13

Chemical Identifiers

132741-29-8
188.13
BKHXODARAOCNDJ-UHFFFAOYSA-N
588647
C1=CC(=C(C=C1C(C(=O)O)O)F)F
C8H6F2O3
MFCD00061299
3,4-difluoromandelic acid, 2-3,4-difluorophenyl-2-hydroxyacetic acid, 3,4-difluorophenyl hydroxy acetic acid, difluoromandelicacid, 3,4-difluoro mandelic acid, 3,4-difluorophenyl hydroxy acetic acid #, alpha-hydroxy-3,4-difluorobenzeneacetic acid, 2-3,4-difluorophenyl-2-hydroxy-acetic acid, 2-3,4-bis fluoranyl phenyl-2-oxidanyl-ethanoic acid
2-(3,4-difluorophenyl)-2-hydroxyacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents