Please login to your online account to display your discounted pricing

3,4-Difluoroiodobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Difluoroiodobenzene
Name Note 99%
CAS 64248-58-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3F2I
Linear Formula IC6H3F2
Formula Weight 239.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC233180250

  SDS
thermo fisher scientific
233180250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 239.99
Quantity 25g
Color Clear colorless to pink or slightly brown
TSCA TSCA
Density 1.9900g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Refractive Index 1.5570 to 1.5590 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 64248-58-4
Linear Formula IC6H3F2
MDL Number MFCD00044630
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H3F2I
Chemical Name or Material 3, 4-Difluoroiodobenzene
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.