Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorocinnamic acid, 97%

$44.45 - $135.92

Chemical Identifiers

Chemical Name or Material 3,4-Difluorocinnamic acid
CAS 112897-97-9
Molecular Formula C9H6F2O2
Formula Weight 184.14
MDL Number MFCD00010320
Beilstein 7370322
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2023003

  SDS
alfa aesar™
B2023003
1g Each for $44.45

AAB2023006

  SDS
alfa aesar™
B2023006
5g Each for $135.92
Description & Specifications

Specifications

Formula Weight 184.14
Chemical Name or Material 3,4-Difluorocinnamic acid
Assay 97%
Molecular Formula C9H6F2O2
MDL Number MFCD00010320
CAS 112897-97-9
Beilstein 7370322
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 194° to 196°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.