Please login to your online account to display your discounted pricing

3,4-Difluorobenzylamine, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  311530010

 View more versions of this product

Catalog No. AC311530010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 143.14
Quantity 1g
Color Clear colorless to yellow
Density 1.2100g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3, 4-Difluorobenzylamine
Storage Information May darken during storage
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 95%
Molecular Formula C7H7F2N
Linear Formula F2C6H3CH2NH2
MDL Number MFCD00010145
CAS 72235-53-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)

Specifications

Chemical Name or Material 3, 4-Difluorobenzylamine
Name Note 97%
CAS 72235-53-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H7F2N
Linear Formula F2C6H3CH2NH2
Formula Weight 143.14
MDL Number MFCD00010145
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents